Learn More
Biomatik Corporation Pig S100 Calcium Binding Protein A8 (S100A8) ELISA Kit
Supplier:  Biomatik Corporation EKU12489
Encompass_Preferred
Pig S100 Calcium Binding Protein A8 (S100A8) ELISA Kit for detecting S100A8 in serum, plasma, tissue homogenates, cell lysates, cell culture supernates, other biological fluids. Species Reactivity: Pig (Sus scrofa; Porcine). Detection Range: 15.6-1,000pg/ml. Sensitivity: 5.2pg/ml. Format: 96-Well Plate. Assay Type: Double-antibody Sandwich. Detection Method: Colormetric. Assay Time: 3h. Accession Number: C3S7K5. Gene ID: 100127488. Target Synonyms: MRP8; MIF; 60B8AG; CAGA; CFAG; CGLA; CP-10; L1Ag; NIF; P8; Calgranulin A; Cystic fibrosis antigen; Leukocyte L1 complex light chain; Migration inhibitory factor-related protein 8. Restrictions: For Research Use Only. Not for use in diagnostic procedures.
Catalog No. 50-241-7598
Only null left
Add to Cart
Product Content Correction
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.
Product Title
Your feedback has been submitted: Thank you for helping us improve our website.