Learn More
New Pig PIG™ Spill Kit in High-Visibility Mobile Container
SDP
Catalog No. 18389119 Shop All New Pig Corporation Products
Change view
Click to view available options
Type: wheeled container
Catalog No.
18-389-119 Type: wheeled container
1 options
This item is not returnable. View return policy
Catalog No. 18-389-119 Supplier New Pig Supplier No. KIT273
Add to Cart
Add to Cart
This item is not returnable. View return policy
Lightweight high-visibility wheeled container kit rolls to spills of up to 37 gallons.
  • Lightweight, 100% polyethylene container resists chemicals Heavy-duty 8 in. rubber wheels roll easily over nearly any surface
  • PIG Blue Socks stop spreading spills;
  • Prepacked container provides easy access to contents
  • Built-in handles allow easy steering through tight spots
  • Bold lettering for quick identification
  • Hinged lid provides easy access
  • Temporary disposal bags help make cleanup easier
  • Tamperproof seal helps prevent pilfering of spill response supplies
WARNING: Cancer - www.P65Warnings.ca.gov