Learn More
New Pig PIG™ Spill Station Safety Message Grippy™ Floor Mat
SDP
Catalog No. 18389115 Shop All New Pig Corporation Products
Change view
Click to view available options
Color:
Black
Catalog No. Color
18-389-115 Black
1 options
This item is not returnable. View return policy
Catalog No. 18-389-115 Supplier New Pig Supplier No. GRPWS21709BK
Add to Cart
Add to Cart
This item is not returnable. View return policy
The Spill Station Safety Message Adhesive-Backed Grippy Floor Mat is easy to install, stays down and won't leave residue or damage floors upon removal.
  • PIG Spill Station Safety Message Adhesive-Backed Grippy Floor Mat identifies workplace caution without shifting, bunching or tripping up workers
  • High-visibility, 14 in. safety image makes it easy to locate a spill kit in an emergency situation
  • Nonpermanent adhesive sticks to clean, dry floors with a super-tight grip, but peels up easily without damaging floors or leaving a residue
  • Unlike vinyl decals or tapes, our adhesive backing has been proven in years of commercial and industrial use and is durable enough to withstand heavy foot traffic, forklifts and pallet jacks
  • Anti-slip sign absorbs moisture and won't become slippery when wet like decals, tapes or paints can
  • Leave in place and clean with mop or floor scrubber; can be scrubbed, vacuumed or swept off frequently without affecting performance
  • Low profile mat withstands heavy foot and cart traffic and stays on the job for up to 3–6 months
  • Durable, high-contrast message and images will not wear off
  • Sticks to most commercial floor surfaces including smooth indoor concrete, vinyl, linoleum, ceramic tile, quarry tile and laminate flooring
Specifications
Material Polypropylene
Color Black