Learn More
American Research Products Inc Pig Testosterone T ELISA Kit
Supplier:  American Research Products Inc CSBE06796P
Encompass
Pig Testosterone T ELISA Kit from CUSABIO Detection Range 0 1 ng/mL-20 ng/mL Sensitivity 0 05 ng/mL Sample serum plasma tissue homogenates Method Competitive
Catalog No. 50-003-26285