Learn More
Abcam Pig TNF alpha ELISA Kit, 1 x 96 Tests
Supplier:  Abcam AB1007561X96TESTS
Encompass_Preferred
Pig TNF alpha ELISA Kit is a Sandwich (quantitative) ELISA kit for the measurement of Pig TNF alpha in Pig in Plasma, Cell culture supernatant, Serum samples.
The product is subject to the following: Abcam Restricted Use Statement
Catalog No. 50-258-5676
Only null left
Add to Cart
Product Content Correction
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.
Product Title
Your feedback has been submitted: Thank you for helping us improve our website.