Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

New Pig PIG Ultra-Duty Adhesive-Backed Grippy Floor Mat
SDP

Catalog No. 17987515 Shop All New Pig Corporation Products
Change view
Click to view available options
Dimensions (L x W):
3 x 3 ft.
5 x 3 ft.
Color:
Black
Gray
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Dimensions (L x W) Color
17-987-515 3 x 3 ft. Gray
17-987-517 5 x 3 ft. Black
17-987-518 5 x 3 ft. Gray
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Catalog No. 17-987-515 Supplier New Pig Supplier No. GRP36205GY
Add to Cart
Add to Cart

Upgrade to the super-safe, adhesive-backed ultra-duty floor mat.

  • Absorbent and versatile, this adhesive floor mat creates a safe, slip-resistant walking surface in entrances, aisleways, and wet areas like doorways, breakrooms, loading docks, and under fountains or beverage station
  • It absorbs water, oil, and grease, traps dirt and salt, dries quickly, and prevents moisture from reaching the floor—ensuring safety and cleanliness with unmatched performance
  • Roll of 15 precut 3 x 3 ft. mediumweight mats make it easy to install a high-traction walking surface in wet problem spots for customers and employees; replaces outdated rental and rubber-backed floor mats and rugs
  • Place in doorways and entrances, walkways and aisleways, transition areas, breakrooms and cafeterias, along produce walls or floral displays, on production floors, ramps and loading docks, beneath water fountains, ice machines and beverage stations or under liquid hazard areas
  • Adhesive-backed floor mat stays exactly where you put it with no shifting, bunching up or ripples that cause slips and trips
  • Dries out quickly; liquidproof barrier layer prevents moisture from soaking through to the floor
  • Absorbs water, oil and grease and traps dirt and salt to keep it from being tracked around
  • Sticks to the floor with a super-tight grip, but peels up easily without leaving a residue or damaging floors
  • Weighs less than rental and rubber-backed mats for less hassle, saves you up to 50% per year on rental contracts and has a lower environmental impact
  • Easy to clean: you can sweep, vacuum, mop, use your shop/wet vac or run your floor scrubber over Grippy Mat and it won't budge
  • Stays in place for up to 3 to 6 months, depending on conditions and traffic patterns
  • The thinnest commercial floor mat you can buy: no rippled edges to obstruct doors or create tripping hazards
  • ADA compliant for wheelchairs and walkers; carts and pallet jacks roll right over it
  • Sticks to most commercial floor surfaces including concrete, vinyl, linoleum, ceramic tile, quarry tile and laminate flooring
  • Tested and Certified by the National Floor Safety Institute (NFSI): high-traction surfaces such as PIG Grippy Floor Mat have been proven to reduce slip-and-fall claims by as much as 90%

Specifications

Width (English) 3 ft.
Width (Metric) 91 cm
Certifications/Compliance ADA Compliant, NFSI Certified
Dimensions (L x W) 3 x 3 ft.
Length (English) 3 ft.
Length (Metric) 91 cm
Material Polypropylene
Color Gray
Type Adhesive Floor Mat
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.