Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

New Pig PIG Ultra-Duty Adhesive-Backed Grippy Floor Mat Starter Pack
SDP

Catalog No. 17987516 Shop All New Pig Corporation Products
Change view
Click to view available options
Length (English):
25 ft.
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Length (English)
17-987-516 25 ft.
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 17-987-516 Supplier New Pig Supplier No. GRP36206GY
Add to Cart
Add to Cart

Upgrade to the super-safe, adhesive-backed ultra-duty floor mat with this all-in-one pack.

  • Absorbent and versatile, this adhesive floor mat creates a safe, slip-resistant walking surface in entrances, aisleways, and wet areas like doorways, breakrooms, loading docks, and under fountains or beverage stations
  • It absorbs water, oil, and grease, traps dirt and salt, dries quickly, and prevents moisture from reaching the floor—ensuring safety and cleanliness with unmatched performance
  • 3 x 25 ft. mediumweight roll allows you to customize the amount of walk off and floor coverage needed to provide a high-traction walking surface for customers and employees in wet problem spots in your facility; replaces outdated rental and rubber-backed floor mats and rugs
  • Contains a 3 ft. W x 25 ft. L roll of PIG Ultra-Duty Grippy Mat, a Safety Knife and Installation Board
  • Place in doorways and entrances, walkways and aisleways, transition areas, breakrooms and cafeterias, along produce walls or floral displays, on production floors, ramps and loading docks, beneath water fountains, ice machines and beverage stations or under liquid hazard areas
  • Adhesive-backed floor mat stays exactly where you put it with no shifting, bunching up or ripples that cause slips and trips
  • Dries out quickly; liquidproof barrier layer prevents moisture from soaking through to the floor
  • Absorbs water, oil and grease and traps dirt and salt to keep it from being tracked around
  • Sticks to the floor with a super-tight grip, but peels up easily without leaving a residue or damaging floors
  • Weighs less than rental and rubber-backed mats for less hassle, saves you up to 50% per year on rental contracts and has a lower environmental impact
  • Easy to clean: you can sweep, vacuum, mop, use your shop/wet vac or run your floor scrubber over Grippy Mat and it won't budge
  • Stays in place for up to 3 to 6 months, depending on conditions and traffic patterns
  • The thinnest commercial floor mat you can buy: no rippled edges to obstruct doors or create tripping hazards
  • ADA compliant for wheelchairs and walkers; carts and pallet jacks roll right over it
  • Sticks to most commercial floor surfaces including concrete, vinyl, linoleum, ceramic tile, quarry tile and laminate flooring
  • Utility Board and Cutting Guide is a convenient, pre-scored cutting board that prevents damage to floors while it helps you make clean, accurate cuts, perfect corners and 45° angles (included)
  • Safety Cutting Knife with recessed blade and protective edges lets you make quick, clean cuts and avoid injuries (included
  • Tested and Certified by the National Floor Safety Institute (NFSI): when used as part of your floor safety program, high-traction surfaces such as PIG Ultra-Duty Grippy Mat have been proven to reduce slip-and-fall claims by as much as 90%

Specifications

Width (Metric) 91 cm
Width (English) 3 ft.
Type Adhesive Floor Mat
Material Polypropylene
Certifications/Compliance ADA Compliant, NFSI Certified
Color Gray
Length (English) 25 ft.
Length (Metric) 7.6 m
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.