Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

New Pig PIG™ Ultra-Duty Adhesive-Backed Grippy™ Floor Mat
SDP

Catalog No. 18389107 Shop All New Pig Corporation Products
Change view
Click to view available options
Dimensions (L x W):
50 ft. x 3 ft.
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Dimensions (L x W)
18-389-107 50 ft. x 3 ft.
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Catalog No. 18-389-107 Supplier New Pig Supplier No. GRP36201BK
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

Hassling with rubber-backed floor mats or rental mats? Make the switch to the mat that sticks; this super-safe, adhesive-backed ultra-duty floor mat creates a safe walking surface in entrances and aisleways.

  • Place in doorways and entrances, walkways and aisleways, transition areas, breakrooms and cafeterias, along produce walls or floral displays, on production floors, ramps and loading docks, beneath water fountains, ice machines and beverage stations or under liquid hazard areas
  • Adhesive-backed floor mat stays exactly where you put it with no shifting, bunching up or ripples that cause slips and trips
  • Dries out quickly without moisture soaking through to the floor; absorbs water, oil and grease and traps dirt and salt to keep it from being tracked around
  • Sticks to the floor with a super-tight grip, but peels up easily without leaving a residue or damaging floors
  • Weighs less than rental and rubber-backed mats for less hassle, saves you up to 50% per year on rental contracts and has a lower environmental impact
  • The thinnest commercial floor mat you can buy: no rippled edges to obstruct doors or create tripping hazards.
  • ADA compliant for wheelchairs and walkers; carts and pallet jacks roll right over it
  • Sticks to most commercial floor surfaces including concrete, vinyl, linoleum, ceramic tile, quarry tile and laminate flooring
  • Tested and Certified by the National Floor Safety Institute (NFSI): high-traction surfaces such as PIG Ultra-Duty Grippy Mat have been proven to reduce slip-and-fall claims by as much as 90%

Specifications

Dimensions (L x W) 50 ft. x 3 ft.
Certifications/Compliance ADA compliant for wheelchairs and walkers
Quantity One roll
Material Polypropylene
Type Floor Mat
Color Black
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.