Learn More
New Pig PIG™ Urinal Grippy™ Mat
SDP
Catalog No. 18389108 Shop All New Pig Corporation Products
Change view
Click to view available options
Color:
Black
Catalog No. Color
18-389-108 Black
1 options
This item is not returnable. View return policy
Catalog No. 18-389-108 Supplier New Pig Supplier No. GRP7001BK
Add to Cart
Add to Cart
This item is not returnable. View return policy
Keep your restrooms cleaner and provide better drip and splash protection with our adhesive-backed urinal mats.
  • Permanently infused antimicrobial technology prevents the growth of odor-causing bacteria and mold on the mats to keep restrooms cleaner and fresher smelling
  • Proprietary, full-coverage adhesive backing keeps mats in place underfoot with no shifting, curling or flipping up to stop slip and trip hazards; peels up easily
  • Mats stick to your floor and won't budge while you clean them; simply mop them with the rest of the floor
  • Technology captures urine and water on contact, wicks them away and dries out quickly Impermeable backing layers stop liquid pass-through to prevent stains and grout damage
  • Tested and certified by the National Floor Safety Institute (NFSI) as a high-traction surface which can reduce slip and fall claims by as much as 90% when used as part of a Floor Safety Program Release
Specifications
Color Black
Type Urinal Mat
Material Polypropylene