Learn More
Biomatik Corporation Pig Vascular Endothelial Growth Factor Receptor 2 (VEGFR2) ELISA Kit
Supplier:  Biomatik Corporation EKU08108
Encompass_Preferred
Pig Vascular Endothelial Growth Factor Receptor 2 (VEGFR2) ELISA Kit for detecting VEGFR2 in serum, plasma, other biological fluids. Species Reactivity: Pig (Sus scrofa; Porcine). Detection Range: 1.56-100ng/ml. Sensitivity: 0.62ng/ml. Format: 96-Well Plate. Assay Type: Double-antibody Sandwich. Detection Method: Colormetric. Assay Time: 3h. Accession Number: F1RWF1. Target Synonyms: CD309; FLK1; VEGFR; KDR; A Type III Receptor Tyrosine Kinase; Kinase Insert Domain Receptor; Kinase Insert Domain Receptor; Fetal Liver Kinase-1. Restrictions: For Research Use Only. Not for use in diagnostic procedures.
Catalog No. 50-241-5463
Only null left
Add to Cart
Product Content Correction
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.
Product Title
Your feedback has been submitted: Thank you for helping us improve our website.