Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ PIGL Polyclonal Antibody
GREENER_CHOICE

Catalog No. PIPA542879
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PIPA542879 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. PIPA542879 Supplier Invitrogen™ Supplier No. PA542879
Only null left
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

Peptide sequence: MEAMWLLCVA LAVLAWGFLW VWDSSERMKS REQGGRLGAE SRTLLVIAHP Sequence homology: Human: 100%.

This gene encodes an enzyme that catalyzes the second step of glycosylphosphatidylinositol (GPI) biosynthesis, which is the de-N-acetylation of N-acetylglucosaminylphosphatidylinositol (GlcNAc-PI). Study of a similar rat enzyme suggests that this protein localizes to the endoplasmic reticulum.
TRUSTED_SUSTAINABILITY

Specifications

Antigen PIGL
Applications Western Blot
Classification Polyclonal
Concentration 0.5 mg/mL
Conjugate Unconjugated
Formulation PBS with 2% sucrose and 0.09% sodium azide
Gene PIGL
Gene Accession No. Q9Y2B2
Gene Alias CHIME; N-acetylglucosaminylphosphatidylinositol deacetylase; N-acetylglucosaminyl-phosphatidylinositol de-N-acetylase; phosphatidylinositol glycan anchor biosynthesis class L; phosphatidylinositol glycan anchor biosynthesis, class L; phosphatidylinositol glycan, class L; phosphatidylinositol-glycan biosynthesis class L protein; PIGL; PIG-L
Gene Symbols PIGL
Host Species Rabbit
Immunogen Synthetic peptide directed towards the N-terminal of human PIGL (aa 1-50).
Purification Method Affinity Chromatography
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 9487
Target Species Human
Content And Storage -20°C, Avoid Freeze/Thaw Cycles
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
WARNING: Cancer - www.P65Warnings.ca.gov
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.