Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ PIGW Polyclonal Antibody
GREENER_CHOICE

Catalog No. PIPA5104012
Change view
Click to view available options
Quantity:
100 μL
Catalog No. Quantity
PIPA5104012 100 μL
1 options

Catalog No. PIPA5104012

Supplier: Invitrogen™ PA5104012

Only null left
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

Antibody detects endogenous levels of total PIGW.

Phosphatidylinositol-glycans (PIGs) are multi-pass transmembrane proteins that localize to the endoplasmic reticulum. PIGs exhibit various functions, but all are crucial for the biosynthesis of the glycosylphosphatidylinositol (GPI)-anchor. Some PIG proteins are components of the GPI transamidase complex and play a role in the recognition of either the GPI attachment signal or the lipid portion of GPI. Other PIGs belong to the glycosyltransferase complex and function in the transfer of N-acetylglucosamine (GlcNAc) to phosphatidylinositol (PI). A variety of other PIGs play distinct roles in GPI synthesis including mannosylation of the GPI-anchor. PIG-W (Phosphatidylinositol-glycan biosynthesis class W protein) is a 504 amino acid multi-pass membrane protein that functions in the third step of GPI biosynthesis and acylates the inositol ring of phosphatidylinositol.
TRUSTED_SUSTAINABILITY

Specifications

Antigen PIGW
Applications Immunohistochemistry (Paraffin), Western Blot, Immunocytochemistry
Classification Polyclonal
Concentration 1 mg/mL
Conjugate Unconjugated
Formulation PBS with 50% glycerol and 0.02% sodium azide; pH 7.4
Gene PIGW
Gene Accession No. Q7Z7B1
Gene Alias 2610044A17Rik; Gwt1; HPMRS5; phosphatidylinositol glycan anchor biosynthesis class W; phosphatidylinositol glycan anchor biosynthesis, class W; phosphatidylinositol glycan, class W; phosphatidylinositol-glycan biosynthesis class W protein; Pigw; PIG-W
Gene Symbols PIGW
Host Species Rabbit
Immunogen A synthesized peptide derived from human PIGW(Accession Q7Z7B1), corresponding to amino acid residues F132-C182.
Purification Method Affinity Chromatography
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 284098
Target Species Human
Content And Storage -20°C
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
WARNING: Cancer - www.P65Warnings.ca.gov
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.