Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ PIK3CA Monoclonal Antibody (H.843.0)
GREENER_CHOICE

Catalog No. PIMA514870 Shop All Thermo Scientific Products
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PIMA514870 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. PIMA514870 Supplier Invitrogen™ Supplier No. MA514870
  • $725.50
    $550.65 / Each of 1

    Save $174.85

    24% Off The following deviations and disclosures may produce prices below stated discounts but will not trigger price reductions: Price shown reflects a temporary, promotional price reduction. No other purchase or commercial obligation required to receive the reduced price. Price may change at any time. Promotional price valid on web orders only. Your contract pricing may differ. Other exclusions apply.
Add to Cart
Edge
Add to Cart

missing translation for 'validMainframeMsg'

Rabbit Monoclonal Antibody

It is not recommended to aliquot this antibody.

Phosphatidylinositol 3-kinase is composed of an 85 kDa regulatory subunit and a 110 kDa catalytic subunit. The protein encoded by this gene represents the catalytic subunit, which uses ATP to phosphorylate PtdIns, PtdIns4P and PtdIns(4,5)P2. This gene has been found to be oncogenic and has been implicated in cervical cancers.
TRUSTED_SUSTAINABILITY

Specifications

Antigen PIK3CA
Applications Immunoprecipitation, Western Blot
Classification Monoclonal
Clone H.843.0
Concentration 121 μg/mL
Conjugate Unconjugated
Formulation 0.01M HEPES with 50% glycerol, 100 μg/mL BSA, 0.15M NaCl and <0.02% sodium azide, pH 7.5
Gene PIK3CA
Gene Accession No. A0A0G2K344, P32871, P42336, P42337
Gene Alias 2.7.1.153; 6330412C24Rik; caPI3K; CLOVE; CWS5; EC:2.7.11.1; GRB1; MCAP; MCM; MCMTC; MGC142161; MGC142163; p110; p110alpha; p110-alpha; phophatidylinositol 3-kinase; phosphatidylinositol 3-kinase, catalytic, 110-KD, alpha; phosphatidylinositol 3-kinase, catalytic, alpha polypeptide; phosphatidylinositol 4,5-bisphosphate 3-kinase 110 kDa catalytic subunit alpha; Phosphatidylinositol 4,5-bisphosphate 3-kinase catalytic subunit alpha isoform; phosphatidylinositol-4,5-bisphosphate 3-kinase 110 kDa catalytic subunit alpha; phosphatidylinositol-4,5-bisphosphate 3-kinase catalytic subunit alpha; phosphatidylinositol-4,5-bisphosphate 3-kinase catalytic subunit alpha isoform; phosphatidylinositol-4,5-bisphosphate 3-kinase catalytic subunit, alpha isoform; phosphatidylinositol-4,5-bisphosphate 3-kinase, catalytic subunit alpha; phosphoinositide-3-kinase catalytic alpha polypeptide; phosphoinositide-3-kinase, catalytic, alpha polypeptide; PI3K; PI3K alpha; PI3Kalpha; PI3K-alpha; pi3kca; PI3-kinase p110 subunit alpha; PI3-kinase subunit alpha; Pik3ca; PIK3R1; pk3ca; ptdIns-3-kinase p110; ptdIns-3-kinase subunit alpha; ptdIns-3-kinase subunit p110-alpha; Serine/threonine protein kinase PIK3CA
Gene Symbols PIK3CA
Host Species Rabbit
Immunogen Synthetic peptide surrounding Asp520 of the sequence of human PI3K p110a.
Purification Method Affinity chromatography
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 170911, 18706, 282306, 5290
Target Species Bovine, Human, Mouse, Rat
Content And Storage -20°C
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
WARNING: Cancer - www.P65Warnings.ca.gov
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.