Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ Human PIK3CB/PIK3R1 (p110 beta/p85 alpha), His Tag Recombinant Protein

Catalog No. PV6454
Encompass_Preferred
Change view
Click to view available options
Quantity:
20 μg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PV6454 20 μg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. PV6454 Supplier Invitrogen™ Supplier No. PV6454

Recombinant Protein

For maximum recovery please spin prior to use. Unless noted below, aliquots of the 5 μg, 10 μg and 20 μg sizes of kinase are not recommended as materials can be used in original packaging until exhausted. For larger sizes, the number of freeze/thaws may be reduced by preparing aliquots, aliquots below 20 μL are not recommended. Please never store a kinase diluted. If properly stored at -80C, this product is guaranteed for 6 months from date of purchase. PIK3CB/PIK3R1 (p110β/p85α) Recombinant Human Protein is: Available in 3 convenient sizes Validated in our Adapta™ and Kinase Binding assays A full-length protein that can help advance your research when used in validated assays Related assay platforms:LanthaScreen™ Kinase Binding AssayAdapta™ Kinase Activity Assay Protein Form: Full Length, Recombinant, Active The gene accession number for PIK3CB (p110β) is NP_006210 and for PIK3R1 (p85α) is NP_852664.1.

PIK3CB/PIK3R1 (p110β/p85α) is a phosphoinositide-3-kinase (PI3K) comprised of a catalytic subunit, p110β, and a regulatory subunit, p85α, encoded by the PIK3CB and PIK3R1 genes respectively. As with other members of the PI3K family, p110β has important regulatory roles in cellular processes including cell survival, proliferation, and differentiation. It affects growth factor signaling and survival by phosphorylating inositol lipids. PTEN-deficient cancer cells have been observed to depend on p110β signaling.
TRUSTED_SUSTAINABILITY

Specifications

Accession Number P27986, P42338
Concentration See Label
For Use With (Application) Kinase Assay
Formulation 50mM sodium phosphate with 150mM Imidazole, 25% glycerol, 0.1mM PMSF, 300mM NaCl, 0.25mM DTT and no preservative; pH 7
Gene ID (Entrez) 5291, 5295
Molecular Weight (g/mol) 123.7-84.7 kDa
Name Human PIK3CB/PIK3R1 (p110 beta/p85 alpha), His Tag
pH Range 7
Purification Method Purified
Quantity 20 μg
Storage Requirements -80°C, Avoid Freeze/Thaw Cycles
Regulatory Status RUO
Gene Alias PIK3CB; PIK3R1
Common Name PIK3CB/PIK3R1
Gene Symbol PIK3CB, PIK3R1
Kinase Family PI3 Kinase
Product Type Protein
Conjugate Unconjugated
Species Human
Recombinant Recombinant
Protein Tag His-tag
Sequence Full length
Content And Storage -80°C, Avoid Freeze/Thaw Cycles
Shipping Condition Dry Ice
Expression System Baculovirus
Protein Family Kinases & Inhibitors
Protein Form Full Length, Recombinant, Active
Form Liquid
Research Category Signal Transduction, Drug Development, Oncology
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.