Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ Human PIK3CB/PIK3R2 (p110 beta/p85 beta), His Tag Recombinant Protein

Catalog No. A31084
Encompass_Preferred
Change view
Click to view available options
Quantity:
1 mg
100 μg
20 μg
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
A31084 20 μg
A32910 100 μg
A33395 1 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Catalog No. A31084 Supplier Invitrogen™ Supplier No. A31084
Add to Cart
Add to Cart

Recombinant Protein

For maximum recovery please spin prior to use. Unless noted below, aliquots of the 5 μg, 10 μg and 20 μg sizes of kinase are not recommended as materials can be used in original packaging until exhausted. For larger sizes, the number of freeze/thaws may be reduced by preparing aliquots, aliquots below 20 μL are not recommended. Please never store a kinase diluted. If properly stored at -80C, this product is guaranteed for 6 months from date of purchase. This PIK3CB/PIK3R2 (p110 beta/p85 beta) recombinant human protein (full length) was expressed in insect cells. PIK3CB/PIK3R2 (p110 beta/p85 beta) recombinant human protein is also available in our SelectScreen™ Kinase Profiling Service. Validated for use with the Adapta™ assay Available in multiple pack sizes for convenient use Also available in our SelectScreen Kinase Profiling Service Protein Form: Full Length, Recombinant, Active The gene accession number for PIK3CB is NP_006210.1 and for PIK3R2 is NP_852664.1.

PIK3CB/PIK3R2 lipid kinase (p110 beta/p85 beta) or Phosphatidylinositol 3-kinase (PI3K) phosphorylate the 3-prime OH position of the inositol ring of inositol lipids which functions in signaling pathways that regulate cell growth by virtue of their activation in response to various mitogenic stimuli. PI3Ks are composed of a 110-kDa catalytic subunit and a 85-kDa adaptor subunit. PIK3CB plays a critical role in regulating the formation and stability of alpha-2B-beta-3 integrin adhesion bonds, which are necessary for shear force-induced platelet activation. PIK3CB sustains alpha-2B-beta-3 integrin activation and stabilized platelet aggregation by regulating both integrin-dependent calcium flux and Gi activation of RAP1B.
TRUSTED_SUSTAINABILITY

Specifications

Accession Number O00459, P42338
Concentration See Label
For Use With (Application) Kinase Assay
Formulation 50mM sodium phosphate with 0.25mM DTT, 150mM Imidazole, 25% glycerol, 0.1mM PMSF, 300mM NaCl and no preservative; pH 7
Gene ID (Entrez) 5291, 5296
Molecular Weight (g/mol) 123.7-83.5 kDa
Name Human PIK3CB/PIK3R2 (p110 beta/p85 beta), His Tag
pH Range 7
Purification Method Purified
Quantity 20 μg
Storage Requirements -80°C, Avoid Freeze/Thaw Cycles
Regulatory Status RUO
Gene Alias PIK3CB; PIK3R2
Common Name PIK3CB/PIK3R2
Gene Symbol PIK3CB, PIK3R2
Kinase Family PI3 Kinase
Product Type Protein
Conjugate Unconjugated
Species Human
Recombinant Recombinant
Protein Tag His-tag
Sequence Full length
Content And Storage -80°C, Avoid Freeze/Thaw Cycles
Shipping Condition Dry Ice
Expression System Insect cells
Protein Form Full Length, Recombinant, Active
Form Liquid
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.