Learn More
Description
Specifications
Specifications
| Cross Reactivity | Human,Mouse,Rat |
| Product Type | PIK3R1 |
| For Use With (Application) | Qualitative |
| Gene Alias | GRB1, p85, p85-ALPHA |
| Gene ID (Entrez) | 5295/8503 |
| Quantity | 1 Kit |
| Regulatory Status | RUO |
| Storage Requirements | Store at 4°C for six months. |
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.