Learn More
Selleck Chemical LLC YM201636 5mg 371942-69-7
SDP
Supplier:  Selleck Chemical LLC S12195MG
Encompass_Preferred
YM201636 is a selective PIKfyve inhibitor with IC50 of 33 nM, less potent to p110? and insensitive to Fabl (yeast orthologue). YM-201636 suppresses the growth of liver cancer via the induction of autophagy. *For Research & Development use only.  Product is not intended for drug, household, or human consumption.
Catalog No. NC0922876
Only null left
Add to Cart
Product Content Correction
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.
Product Title
Your feedback has been submitted: Thank you for helping us improve our website.