Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Thermo Scientific™ Optical Adhesive Film, for Piko PCR plates

Catalog No. asf0020
Change view
Click to view available options
Quantity:
100 Sheets
400 Sheets
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
ASF0020 400 Sheets
ASF0020100 100 Sheets
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. ASF0020 Supplier Thermo Scientific™ Supplier No. ASF0020
Only null left
Add to Cart
Add to Cart

The exceptionally clear Thermo ScientificPiko PCR/qPCR Film Seal allows high transmission of fluorescence across the range of commonly-used fluorophores. When used with aPiko Thermal CyclerorPikoReal Real-Time PCR System, sample volumes as low as 5μL can be used with minimal evaporative loss. These optical adhesive films are available in the ¼-size format for Piko PCR Plates.

Highlights

  • Optical clarity with minimal auto-fluorescence for real-time PCR applications
  • Tight sealing for sample volumes as low as 5µL
  • Adhesive backing allows easy application without costly sealing devices
  • Cryo-compatible adhesive is effective to –80°C
TRUSTED_SUSTAINABILITY

Specifications

Color Optical
Quantity 400 Sheets
For Use With (Equipment) Piko™ Thermal Cycler, PikoReal™ Real-Time PCR System
Product Type Film, Seal, or Applicator
Product Line Piko
Adhesive Pressure Sensitive Optical Seal
Plate Compatibility Piko Plate
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.