Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Renco Pilgrim™ Powder Free White Nitrile Gloves
SDP

Catalog No. 12888440C
Change view
Click to view available options
Size:
Large
Medium
Small
X-Large
X-Small
5 product options available for selection
Product selection table with 5 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Size
12-888-440C Medium
12-888-440D Large
12-888-440E X-Large
12-888-440A X-Small
12-888-440B Small
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 5 options available.
5 options
This item is not returnable. View return policy
Catalog No. 12-888-440C Supplier Renco Supplier No. R12P333M

Item Discontinued This item has been discontinued by the Fisher Scientific channel. Please to view product availability in your area.

This item is not returnable. View return policy

Nitrile powder free exam gloves incorporate advanced proprietary formula to provide sensitivity, strength and comfort for extended wear.

  • Advanced proprietary low modulus formulation provides premium strength, durability, and sensitivity
  • Superior stretch and comfort for extended wear
  • Polymer coated for ease of donning
  • Excellent dry & wet grip
  • Latex-Free
  • Powder Free
  • Uniform 3 mil thickness
  • Medical Grade
  • 9.5 in. Length
  • White
  • Made in the USA

Specifications

Chemotherapeutic Rated No
Applications Healthcare, Laboratory
AQL 2.5
Color White
Disposable Single-use
Exterior Finish Fully Textured
Latex Free Yes
Material Nitrile
Powdered Powder Free
Product Type Nitrile Examination Glove
Size Medium
Thickness (English) 3 mil
Thickness (Metric) 0.07 mm
FDA 510 (k) Yes
Model R12-P333M
Quantity 1000/Cs.
Series Pilgrim™
Tensile Strength 14 Mpa
Length (English) 9.5 in.
Length (Metric) 24.13 cm
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.