Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Pilocarpine Nitrate, Crystal, USP, 98.5-101%, Spectrum™ Chemical
SDP

Catalog No. 18605219 Shop All Spectrum Chemical Manufacturing Corporation Products
Change view
Click to view available options
Quantity:
1 g
Packaging:
Amber Glass Bottle
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity Packaging
18-605-219 1 g Amber Glass Bottle
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 18-605-219 Supplier Spectrum Chemical Manufacturing Corporation Supplier No. PI1101GM

Item Discontinued This item has been discontinued by the supplier. Please to view product availability in your area.

PI110, 148-72-1, C11H16N2O2·HNO3

Spectrum™ Chemical Pilocarpine Nitrate, Crystal, USP is a medication used to treat glaucoma and dry mouth. All Spectrum Chemical USP grade products are manufactured, packaged and stored under current Good Manufacturing Practices (cGMP) per 21CFR part 211 in FDA registered and inspected facilities.

Chemical Identifiers

CAS 148-72-1
Molecular Formula C11H17N3O5
Molecular Weight (g/mol) 271.27
InChI Key PRZXEPJJHQYOGF-LQRGNCEWSA-N
IUPAC Name (3S)-3-ethyl-4-[(1-methyl-1H-imidazol-5-yl)methyl]oxolan-2-one; nitric acid
SMILES O[N+]([O-])=O.CC[C@H]1C(CC2=CN=CN2C)COC1=O

Specifications

CAS Max % 1
Loss on Drying 0.02
Packaging Amber Glass Bottle
Quantity 1 g
Specific Rotation +79.5° to +82.5°
Percent Purity 98.5 to 101%
Grade USP
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.