Learn More
Heidolph™ Pilot Process Standard Jacketed Vessel Kits
Heidolph™ Pilot Process Standard Jacketed Vessel Kits for use with Reactor-Ready Pilot Lab Reactor
$23984.58
Specifications
Includes | Process Vessel, Turbine Stirrer Paddle, PTFE Pt100 Temperature Probe with Lemo Plug, PTFE Temperature Probe Adapter |
---|---|
Capacity (Metric) | 20 L |
For Use With (Equipment) | Reactor-Ready Pilot Lab Reactor |
Catalog Number | Mfr. No. | Type | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Catalog Number | Mfr. No. | Type | Price | Quantity & Availability | |||||
13-890-023
![]() |
Heidolph™
015908016 |
Single Jacketed |
Each of 1 for $23,984.58
|
|
|||||
Description
- Vessels have 1.25 to 1 ratio of internal height to diameter to mimic plant scale reactors
- DN200 vessel flange
Specifications
Process Vessel, Turbine Stirrer Paddle, PTFE Pt100 Temperature Probe with Lemo Plug, PTFE Temperature Probe Adapter | |
20 L | |
Reactor-Ready Pilot Lab Reactor |