Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ PIM1 Polyclonal Antibody
GREENER_CHOICE

Catalog No. PIPA587366
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PIPA587366 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. PIPA587366 Supplier Invitrogen™ Supplier No. PA587366
Only null left
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

Immunogen sequence: MLLSKINSLA HLRAAPCNDL HATKLAPGKE KEPLESQYQV GPLLGSGGFG SVYSGIRVSD NLPVAIKHVE KDRISDWGEL PNGTRVPMEV VLLKKVSSGF SGVIRLLDWF ERPDSFVLIL ERPEPVQDLF DFITERGALQ EELARSFFWQ VLEAVRHCHN CGVLHRDIKD ENILIDLNRG ELKLIDFGSG ALLKDTVYTD FDGTRVYSPP EWIRYHRYHG RSAAVWSLGI LLYDMVCGDI PFEHDEEIIR GQVFFRQRVS SECQHLIRWC LALRPSDRPT FEEIQNHPWM QDVLLPQETA EIHLHSLSPG PSK; Positive Samples: Rat kidney; Cellular Location: Cell membrane, Cytoplasm, Nucleus.

The proto-oncogene PIM1 is upregulated in prostate cancer. PIM1, which belongs to the Serine/Threonine protein kinase family, is thought to play a role in signal transduction in blood cells. The protooncogene PIM1 encodes a protein kinase upregulated in prostate cancer. It may affect the structure or silencing of chromatin by phosphorylating HP1 gamma/CBX3. PIM1 is expressed primarily in cells of the hematopoietic and germ line lineages.
TRUSTED_SUSTAINABILITY

Specifications

Antigen PIM1
Applications ELISA, Western Blot
Classification Polyclonal
Concentration 1.90 mg/mL
Conjugate Unconjugated
Formulation PBS with 50% glycerol and 0.01% thimerosal; pH 7.3
Gene PIM1
Gene Accession No. P26794
Gene Alias non-specific serine/threonine protein kinase; Oncogene PIM1; PIM; Pim1; Pim-1; pim-1 kinase 44 kDa isoform; pim-1 oncogene; pim-1 oncogene (proviral integration site 1); Pim-1 proto-oncogene, serine/threonine kinase; proto-oncogene serine/threonine-protein kinase Pim-1; proviral integration site 1; serine/threonine-protein kinase pim-1; serine-threonine kinase PIM-1
Gene Symbols PIM1
Host Species Rabbit
Immunogen Recombinant fusion protein containing a sequence corresponding to amino acids 1-313 of human PIM1 (NP_0026391).
Purification Method Affinity Chromatography
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 24649
Target Species Rat
Content And Storage -20°C, Avoid Freeze/Thaw Cycles
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.