Learn More
TARGETMOL CHEMICALS INC Laromustine 1MG
SDP
Supplier:  TARGETMOL CHEMICALS INC T702291MG
Encompass_Preferred
Also available in 5 mg, 10 mg, 25 mg, 50 mg and bulk. Please contact Fisher for quotes. Laromustine (VNP40101M) is a sulfonyl hydrolytic alkylating prodrug used in cancer therapy with significant anticancer activity. Inhibiting thioredoxin reductase can be used to study acute myeloid leukemia.
Catalog No. 50-287-7838
Only null left
Add to Cart
Product Content Correction
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.
Product Title
Your feedback has been submitted: Thank you for helping us improve our website.