Learn More
Selleck Chemical LLC PF-06447475
SDP
Supplier:  Selleck Chemical LLC S820210MG
Encompass_Preferred
PF-06447475 is a potent selective and brain penetrant LRRK2 kinase inhibitor with IC50 of 3 nM
Catalog No. 50-000-01751
Only null left
Add to Cart
Product Content Correction
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.
Product Title
Your feedback has been submitted: Thank you for helping us improve our website.