Learn More
Selleck Chemical LLC Pimecrolimus S5004-50mg
SDP
Supplier:  Selleck Chemical LLC S500450MG
Encompass_Preferred
Pimecrolimus (Elidel ASM 981 SDZ-ASM 981) is an immunophilin ligand which binds specifically to the cytosolic receptor immunophilin macrophilin-12 (FKBP-12) a calcineurin inhibitor
Catalog No. 50-311-0189
Only null left
Add to Cart
Product Content Correction
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.
Product Title
Your feedback has been submitted: Thank you for helping us improve our website.