Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ Pimivalimab Recombinant Monoclonal Antibody

Catalog No. MA559724
Encompass_Preferred
Change view
Click to view available options
Quantity:
1 mg
100 μg
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
MA559724 1 mg
MA559723 100 μg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. MA559724 Supplier Invitrogen™ Supplier No. MA559724
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

Recombinant Monoclonal Antibody

For reconstitution, add sterile, distilled water to achieve a final antibody concentration of 1 mg/mL. Gently shake to solubilize the protein completely. Do not vortex. Reconstituted products should be stored at -80 °.

Pimivalimab, also known as JTX-4014, is a promising anti-PD-1 inhibitor primarily employed in oncology treatments. This biosimilar, located in the immunoglobulin superfamily (IgSF), is strategically designed to target the PD-1 receptor found on T-cells. The gene PDCD1, also known as PD-1 or CD279, encodes Programmed Cell Death Protein 1, a crucial immune checkpoint receptor expressed on T cells and other immune cells. Located on chromosome 2q37.3, PDCD1 consists of five exons. PD-1 is a type I transmembrane protein within the CD28/CTLA-4 superfamily and functions primarily to inhibit immune responses, thus maintaining peripheral tolerance and preventing autoimmune reactions. Upon binding to its ligands, PD-L1 and PD-L2, expressed on tumor cells and other tissues, PD-1 mediates the suppression of T-cell activity, inducing apoptosis in antigen-specific T cells while sparing regulatory T cells, contributing significantly to immune evasion by tumors.
TRUSTED_SUSTAINABILITY

Specifications

Antigen Pimivalimab
Applications ELISA, Flow Cytometry, Functional Assay, Surface Plasmon Resonance
Classification Recombinant Monoclonal
Concentration 1 mg/mL
Conjugate Unconjugated
Formulation 25mM histidine with 8% sucrose, 0.01% Tween 80 and no preservative; pH 6.2
Host Species Human
Purification Method Protein A
Quantity 1 mg
Regulatory Status RUO
Primary or Secondary Primary
Target Species Human
Content And Storage -20°C, Avoid Freeze/Thaw Cycles
Product Type Antibody
Form Lyophilized
Isotype IgG4SP
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.