Learn More
Cayman Chemical ANTIBODY MItoquInol 25mg
SDP
Supplier:  Cayman Chemical 8995025 MG
Encompass_Preferred
A xanthone with anti-inflammatory and enzyme inhibitory activities inhibits NF-KB p65 binding to DNA in a cell-free assay in an enzyme assay (IC50 11.3 UM) inhibits BChE and AChE (IC50s 2.46 and 7.75 UG/ml)
Catalog No. 50-289-2770
Only null left
Add to Cart
Product Content Correction
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.
Product Title
Your feedback has been submitted: Thank you for helping us improve our website.