Learn More
Apexbio Technology LLC Pimozide 2062-78-4 100mg
SDP
Supplier:  Apexbio Technology LLC C3837100
Encompass_Preferred
Pimozide (CAS 2062-78-4) is a small-molecule antagonist targeting dopamine receptors including D2 D3 and D4 subtypes It is designed to inhibit dopamine receptor signaling thereby modulating dopaminergic pathways Pimozide exerts its biological activity primarily through antagonistic binding to dopamine receptors It demonstrates inhibition with Ki values of approximately 2 4 nM (D2) 0 2 nM (D3) and 1 8 nM (D4) In animal studies administration of pimozide reduced dopamine-driven behaviors such as food-motivated activity and cocaine reinforcement Based on these pharmacological properties pimozide holds research potential in neurological and psychiatric disorders including schizophrenia Tourette s syndrome dopamine-related motor disorders and delusional psychosis
Catalog No. 50-000-89127
Only null left
Add to Cart
Product Content Correction
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.
Product Title
Your feedback has been submitted: Thank you for helping us improve our website.