Learn More
PIMT Rabbit anti-Human, Polyclonal, Bethyl Laboratories
Rabbit Polyclonal Antibody
Supplier: Bethyl Laboratories, Inc A300814AT
Description
The recommended shelf life for this product is 1 year from date of receipt. Application Note: For IHC, epitope retrieval with Tris-EDTA pH 9.0 is recommended for FFPE tissue sections.
The nuclear receptor coactivators participate in the transcriptional activation of specific genes by nuclear receptors. Using a coactivator peroxisome proliferator-activated receptor-interacting protein (PRIP) as bait, a novel, protein has been identified, designated PRIP-interacting protein with methyltransferase domain (PIMT). It is expressed in heart, skeletal muscle, kidney, liver, and placenta and is a component of nuclear receptor signal transduction apparatus acting through PRIP.Specifications
| PIMT | |
| Polyclonal | |
| Unconjugated | |
| TGS1 | |
| HCA137, Hepatocellular carcinoma associated antigen 137, NCOA6IP, Nuclear receptor coactivator 6 interacting protein, PIPMT, PRIP interacting protein | |
| Rabbit | |
| Antigen affinity chromatography | |
| RUO | |
| 96764 | |
| 4° C | |
| Liquid |
| Immunohistochemistry, Immunoprecipitation | |
| 0.2 mg/ml | |
| TBS with 0.1% BSA and 0.09% sodium azide | |
| Q96RS0 | |
| TGS1 | |
| Between 150 and 200 | |
| 10 μL | |
| Primary | |
| Human | |
| Antibody | |
| IgG |
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.