Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

PIMT Rabbit anti-Human, Polyclonal, Bethyl Laboratories

Catalog No. 501558515
Encompass_Preferred
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
50-155-8515 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 50-155-8515 Supplier Bethyl Laboratories, Inc Supplier No. A300814A
Only null left
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

The recommended shelf life for this product is 1 year from date of receipt. Application Note: For IHC, epitope retrieval with Tris-EDTA pH 9.0 is recommended for FFPE tissue sections.

The nuclear receptor coactivators participate in the transcriptional activation of specific genes by nuclear receptors. Using a coactivator peroxisome proliferator-activated receptor-interacting protein (PRIP) as bait, a novel, protein has been identified, designated PRIP-interacting protein with methyltransferase domain (PIMT). It is expressed in heart, skeletal muscle, kidney, liver, and placenta and is a component of nuclear receptor signal transduction apparatus acting through PRIP.

Specifications

Antigen PIMT
Applications Immunohistochemistry, Immunoprecipitation
Classification Polyclonal
Concentration 0.2 mg/ml
Conjugate Unconjugated
Formulation TBS with 0.1% BSA and 0.09% sodium azide
Gene TGS1
Gene Accession No. Q96RS0
Gene Alias HCA137, Hepatocellular carcinoma associated antigen 137, NCOA6IP, Nuclear receptor coactivator 6 interacting protein, PIPMT, PRIP interacting protein
Gene Symbols TGS1
Host Species Rabbit
Immunogen Between 150 and 200
Purification Method Antigen affinity chromatography
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 96764
Target Species Human
Content And Storage 4° C
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.