Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

PIN-1 Monoclonal Antibody (ZP004), Abbomax

Catalog No. 502204421
Encompass_Preferred
Change view
Click to view available options
Quantity:
100 μg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
50-220-4421 100 μg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 50-220-4421 Supplier Abbomax Inc Supplier No. 606170
Only null left
Add to Cart
Add to Cart

Mouse Monoclonal Antibody

Positive control: fetal kidney lysate Cellular location: Nucleus.

PIN-1 Monoclonal antibody specifically detects PIN-1 in Human samples. It is validated for Immunohistochemistry (Paraffin), Western Blot

Specifications

Antigen PIN-1
Applications Immunohistochemistry (Paraffin), Western Blot
Classification Monoclonal
Clone ZP004
Concentration 0.5 mg/mL
Conjugate Unconjugated
Formulation PBS with proprietary stabilizer and 0.01% sodium azide
Gene Pin1
Gene Accession No. Q13526
Gene Alias 0610025L01Rik; D9Bwg1161e; DOD; EC 5.2.1.8; NIMA-interacting; peptidylprolyl cis/trans isomerase, NIMA-interacting 1; peptidylprolyl cis/trans isomerase, NIMA-interacting 1 a; peptidylprolyl cis/trans isomerase, NIMA-interacting 1 S homeolog; Peptidyl-prolyl cis-trans isomerase NIMA-interacting 1; peptidyl-prolyl cis-trans isomerase Pin1; pin1; pin1.S; pin1a; pin1-a; pin1-b; PPIase Pin1; prolyl isomerase; prolyl isomerase Pin1; protein (peptidyl-prolyl cis/trans isomerase) NIMA-interacting 1; protein (peptidylprolyl cis/trans isomerase) NIMA-interacting 1; rotamase Pin1; ubl5; XELAEV_18021616mg
Host Species Mouse
Immunogen A recombinant protein PIN-1
Purification Method Protein A
Quantity 100 μg
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 5300
Target Species Human
Content And Storage Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles.
Product Type Antibody
Form Liquid
Isotype IgG2b
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.