Learn More
Medchemexpress LLC PIN1 inhibitor API-1 10mM 1mL | 680622-70-2 | 366.30 g/mol | C15H13F3N6O2 | 1 ML
Supplier: Medchemexpress LLC HY11671610MM 1ML
PIN1 inhibitor API-1 is a small-molecule inhibitor of peptidyl-prolyl cis-trans isomerase NIMA-interacting 1 (Pin1) supplied for research use. The compound is available as a solid and as ready-to-use 10 mM solution in DMSO, and is provided at high reported purity to support reproducible in vitro studies.
- Specific Pin1 inhibitor suitable for cellular and biochemical assays.
- Available as 10 mM solution in DMSO or as solid for custom formulations.
- High purity supports reproducible biological testing.
- Molecular weight 366.30, molecular formula C15H13F3N6O2.
- Supplied in small-volume formats appropriate for screening and dose-response studies.
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.