Learn More
Selleck Chemical LLC PIN1 inhibitor API-1 5mg
SDP
Supplier:  Selleck Chemical LLC S68935MG
Encompass_Preferred
PIN1 inhibitor API-1 is a specificPin1 (peptidyl-prolyl cis-trans isomerase NIMA-interacting 1) inhibitor with IC50 of 72.3 nM. PIN1 inhibitor API-1 targets Pin1 peptidyl-prolyl isomerase domain and inhibits Pin1 cis-trans isomerizing activity. PIN1 inhibitor API-1 up-regulatesmiRNA biogenesis by retaining active XPO5 conformation and suppresses HCC development. *For Research & Development use only.  Product is not intended for drug, household, or human consumption.
Catalog No. 50-217-3232
Only null left
Add to Cart
Product Content Correction
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.
Product Title
Your feedback has been submitted: Thank you for helping us improve our website.