Learn More
Invitrogen™ PIN4 Polyclonal Antibody

Description
Positive Samples: 293T, HepG2, U-251MG, Mouse liver, Mouse brain, Rat testis, Rat brain Immunogen sequence: MPPKGKSGSG KAGKGGAASG SDSADKKAQG PKGGGNAVKV RHILCEKHGK IMEAMEKLKS GMRFNEVAAQ YSEDKARQGG DLGWMTRGSM VGPFQEAAFA LPVSGMDKPV FTDPPVKTKF GYHIIMVEGR K.
Specifications
Specifications
| Antigen | PIN4 |
| Applications | ELISA, Western Blot |
| Classification | Polyclonal |
| Concentration | 3.8 mg/mL |
| Conjugate | Unconjugated |
| Formulation | PBS with 50% glycerol and 0.01% thimerosal; pH 7.3 |
| Gene | Pin4 |
| Gene Accession No. | Q9CWW6, Q9Y237 |
| Gene Alias | 2410002I22Rik; EPVH; eukaryotic parvulin homolog; hEPVH; hPar14; hPar17; MGC138486; Par14; Par17; parvulin; parvulin-14; Parvulin-17; peptidyl-prolyl cis/trans isomerase EPVH; peptidylprolyl cis/trans isomerase NIMA-interacting, 4; peptidylprolyl cis/trans isomerase, NIMA-interacting 4; peptidyl-prolyl cis-trans isomerase NIMA-interacting 4; Peptidyl-prolyl cis-trans isomerase Pin4; PIN4; PPIase Pin4; protein (peptidyl-prolyl cis/trans isomerase) NIMA-interacting, 4 (parvulin); protein (peptidylprolyl cis/trans isomerase) NIMA-interacting, 4 (parvulin); rotamase PIN4 |
| Gene Symbols | Pin4 |
| Show More |
Safety and Handling
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.