Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ PIN4 Polyclonal Antibody
GREENER_CHOICE

Catalog No. PA551206
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PA551206 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. PA551206 Supplier Invitrogen™ Supplier No. PA551206
Only null left
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

The antibody detects endogenous levels of total PIN4 protein.

This gene encodes a member of the parvulin subfamily of the peptidyl-prolyl cis/trans isomerase protein family. The encoded protein catalyzes the isomerization of peptidylprolyl bonds, and may play a role in the cell cycle, chromatin remodeling.
TRUSTED_SUSTAINABILITY

Specifications

Antigen PIN4
Applications Immunohistochemistry, Western Blot
Classification Polyclonal
Concentration 1 mg/mL
Conjugate Unconjugated
Formulation PBS with 40% glycerol and 0.05% sodium azide; pH 7.4
Gene Pin4
Gene Accession No. Q9Y237
Gene Alias 2410002I22Rik; EPVH; eukaryotic parvulin homolog; hEPVH; hPar14; hPar17; MGC138486; Par14; Par17; parvulin; parvulin-14; Parvulin-17; peptidyl-prolyl cis/trans isomerase EPVH; peptidylprolyl cis/trans isomerase NIMA-interacting, 4; peptidylprolyl cis/trans isomerase, NIMA-interacting 4; peptidyl-prolyl cis-trans isomerase NIMA-interacting 4; Peptidyl-prolyl cis-trans isomerase Pin4; PIN4; PPIase Pin4; protein (peptidyl-prolyl cis/trans isomerase) NIMA-interacting, 4 (parvulin); protein (peptidylprolyl cis/trans isomerase) NIMA-interacting, 4 (parvulin); rotamase PIN4
Gene Symbols Pin4
Host Species Rabbit
Immunogen Full length fusion protein of human PIN4.
Purification Method Antigen affinity chromatography
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 5303
Target Species Human
Content And Storage -20°C
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
WARNING: Cancer - www.P65Warnings.ca.gov
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.