Learn More
Eisco™ Pinhole Camera Kit
SCI_ED
Catalog No. S15125 Shop All Eisco Products
Change view
Click to view available options
Pinhole Camera
Catalog No.
S15125 Pinhole Camera
1 options
Catalog No. S15125 Supplier Eisco™ Supplier No. PH0607
Add to Cart
Add to Cart
The Pinhole Camera Kit can be used to investigate the action of a lens, and the construction of a pinhole camera.
  • A great teaching tool to demonstrate how a camera sees things in much the same way as the eye does
  • Proves that large apertures produce brighter images but shorter depths of field
  • Designed for use with a camera approximately 150 mm from front to back
  • Lens used should have a focal length equal to the length of the box
  • Product does not take actual photos, additional camera equipment is required
  • Requires an external power supply (not included)
Includes
  • 8 Pinhole Camera Boxes with Holes (15 x 10 x 10 cm), Screen with Lid, 24 W Filament Lamp, 50 Sheets of Black Paper (20 x 25 cm), 60 W Lamp, 12 V AC/DC Mounted Lamp Holder, and a Packet of Pins
Specifications
Product Type Pinhole Camera
For Use With (Application) Demonstrating How a Camera Works
For Use With (Equipment) Camera (150 mm From Front to Back)
Includes 8 Pinhole Camera Boxes with Holes, Screen with Lid, 24 W Filament Lamp, 50 Sheets of Black Paper, 60 W Lamp, 12 V AC/DC Mounted Lamp Holder, and a Packet of Pins
Dimensions (L x W) 20 x 25 cm (Paper)
Dimensions (L x W x H) 15 x 10 x 10 cm (Camera Box)