Learn More
United Scientific™ PinPoint™ Pipette Controller Nose Cone
SDPSCI_ED
Catalog No. S012426 Shop All United Scientific Supplies, Inc Products
Change view
Click to view available options
Pinpoint Controller, Type: Nose Cone
Catalog No.
S012426 Pinpoint Controller, Type: Nose Cone
1 options
Catalog No. S012426 Supplier United Scientific™ Supplier No. UNPINPNT03
Add to Cart
Add to Cart
Nose cone for use with the PinPoint™ Pipette Controller.
  • Ultra light and powerful pipette controller is designed for comfort as well as accuracy
  • Intuitive 6 level speed setting and sensitive triggers provide a wide range of aspirating/dispense speeds
  • Pipette controller is ergonomically designed for extended usage
  • Compatible with glass or plastic serological pipettes (0.1 to 100 mL)
  • LCD indicates battery charge and 6 speed levels
  • Fills a 25 mL pipette in less than 5 seconds
  • Lithium-ion battery charges fully within 3 hours and can provide up to 8 hours use
  • Meets the requirements of the applicable EU directives, UK regulations, UL, and CAN/CSA
Specifications
For Use With PinPoint Pipette Controller
Type Nose Cone