Learn More
Enzo Life Sciences Pioglitazone (5mg). CAS: 111025-46-8
SDP
Supplier:  Enzo Life Sciences ALX270367M005
Encompass_Preferred
Pioglitazone selectively activates PPARγ-1. It is about one tenth as potent as rosiglitazone (EC50~500nM for human and mouse PPARγ). In a transactivation assay using COS-1 cells transfected with full length human PPARα and RXRα, pioglitazone and rosiglitazone exhibit low level activation of PPARα at 1µM and 5.4- and 4.2-fold activation, respectively, at a concentration of 10µM. Formula: C19H20N2O3S. MW: 356.4. MI: 14: 7452. Purity: ≥97% (HPLC). Appearance: White to off-white solid. Solubility: Soluble in DMSO (2.5mg/ml) or dimethyl formamide (2.5mg/ml). Long Term Storage: -20°C. Handling: Protect from moisture.
Catalog No. 50-200-8513
Only null left
Add to Cart
Product Content Correction
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.
Product Title
Your feedback has been submitted: Thank you for helping us improve our website.