Learn More
Selleck Chemical LLC Pioglitazone HCl 10mg 112529-15-4 AD-4833, U-72107E
SDP
Supplier:  Selleck Chemical LLC S204610MG
Encompass_Preferred
Pioglitazone HCl (AD-4833, U-72107E) is an inhibitor of cytochrome P450 (CYP)2C8 and CYP3A4 enzymes. Pioglitazone HCl inhibits CYP2C8, CYP3A4 and CYP2C9 with Ki of 1.7 ?M, 11.8 ?M and 32.1 ?M, respectively. Pioglitazone HCl is also a selective peroxisome proliferator-activated receptor-gamma (PPAR?) agonist with EC50 of 0.93 ?M and 0.99 ?M for human PPAR? and mouse PPAR?, respectively. Pioglitazone HCl inhibits mitochondrial iron uptake, lipid peroxidation, and subsequent ferroptosis. *For Research & Development use only.  Product is not intended for drug, household, or human consumption.
Catalog No. 50-136-1937
Only null left
Add to Cart
Product Content Correction
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.
Product Title
Your feedback has been submitted: Thank you for helping us improve our website.