Learn More
PIP™ Adversary Safety Eyewear
Catalog No. 19825281 Shop All Protective Industrial Products Inc. Products
Change view
Click to view available options
Lens Tint:
Gray
Light Blue
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Lens Tint
19-825-281 Light Blue
19-148-450 Gray
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Catalog No. 19-825-281 Supplier PIP™ Supplier No. 250280003
Add to Cart
Add to Cart
This item is not returnable. View return policy
Lightweight design provides comfortable fit
  • Semi-rimless, glossy, black rubber temples, one piece bridge frame
  • Soft nose piece
  • Enclosed lanyard
  • Polycarbonate lens with scratch resistant hard coating
  • Base curve: 9
  • Sleek design with 8-base lens curve
Compliance
Exceeds ANSI Z87.1+ High Impact Requirements.
Specifications
Lens Coating Scratch-resistant
Lens Tint Light Blue
Product Type Safety Goggles
Frame Color Black
Material Polycarbonate
Disposable Reusable
Lens Material Polycarbonate
Frame Material Plastic
Frame Type Semi-rimless
Adhere to
Safety Regulations and Best Practices
Take advantage of a range of Safety Services from our trusted suppliers to help ensure the safety and compliance of your people, processes, products, and facilities. Services include assessments, training, equipment maintenance, safety programs, and more.