Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
PIP™ ArmorGrip™ Nitrile Coated Terry Cloth Gloves with Textured Palm
Shop All Protective Industrial Products Inc. ProductsDescription
- Protects hands from physical hazards such as cuts, punctures, snags and abrasions
- Terry cloth palm coated with rough textured blue nitrile
- ActiFresh™ treated to reduce germs and odors
- Safety cuff helps protect the wrist and allows for fast removal
- Heavy-duty protection without sacrificing flexibility or tactile sensitivity
Specifications
Specifications
| Cuff Material | Plasticized |
| Coating Material | Nitrile |
| Color | Blue |
| Exterior Finish | Partially Textured |
| Hand | Specific |
| Product Type | Dip-coated Gloves |
| Size | Large |
| For Use With (Application) | General Assembly, Fabrication, Parts Handling, and Maintenance |
| Glove Cuff Color | Natural |
| Glove Cuff Style | Safety |
| Show More |
Product Title
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.
Spot an opportunity for improvement?