Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

PIP™ Assurance™ Neoprene/Latex Flock-Lined Gloves with Honeycomb Grip

Catalog No. 17730108 Shop All Protective Industrial Products Inc. Products
Change view
Click to view available options
Size:
Small
Medium
Large
X-Large
2X-Large
5 product options available for selection
Product selection table with 5 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Size
17-730-108 2X-Large
17-730-104 Small
17-730-105 Medium
17-730-106 Large
17-730-107 X-Large
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 5 options available.
5 options
Catalog No. 17-730-108 Supplier PIP™ Supplier No. 523671/XXL
Add to Cart
Add to Cart

Unsupported neoprene/latex gloves designed for reliable chemical protection, enhanced grip, and comfortable extended-use wear.

  • Neoprene gloves provide superior protection from a wide range of chemicals such as acids, caustics, solvents and oils
  • Double-dipped so you get the versatile chemical resistance properties of neoprene and the comfort of latex
  • Straight cuff adds additional length to protect the forearm from liquid run-off
  • Flock lining provides increased comfort and ease of donning
  • Honeycomb pattern for an enhanced grip
  • Made from component materials that comply with Federal regulations for food contact 21 CFR, 170-199

Specifications

ANSI Abrasion Rating ANSI 0
Chlorinated No
Applications Industrial, Laboratory
Color Blue, Yellow
Latex Free No
Material Neoprene, Latex
Product Type Chemical-resistant Gloves
Size 2X-Large
Thickness (English) 28 mil
EN 421 No
Grip Design Honeycomb
Lining Flock Lined
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.