Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

PIP™ Assurance™ Unsupported Neoprene Gloves

Catalog No. 19120352 Shop All Protective Industrial Products Inc. Products
Change view
Click to view available options
Size:
2X-Large
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Size
19-120-352 2X-Large
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Catalog No. 19-120-352 Supplier PIP™ Supplier No. 523670/XXL
  • $1,300.00
    $650.65 / Case of 144

    Save $649.35

    50% Off The following deviations and disclosures may produce prices below stated discounts but will not trigger price reductions: Price shown reflects a temporary, promotional price reduction. No other purchase or commercial obligation required to receive the reduced price. Price may change at any time. Promotional price valid on web orders only. Your contract pricing may differ. Other exclusions apply.
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

Ideal for use in the petrochemical industry, janitorial, general maintenance and photo finishing.

  • Neoprene gloves provide superior protection from a wide range of chemicals such as acids, caustics, solvents and oils
  • Double dipped so you get the versatile chemical resistance properties of neoprene and the comfort of latex
  • Straight cuff adds additional length to protect the forearm from liquid run-off
  • Flock lining provides increased comfort and ease of donning
  • Raised Diamond pattern for an enhanced grip
  • Made from component materials that comply with Federal regulations for food contact 21 CFR, 170-199

Specifications

Certifications/Compliance 21 CFR 170-199, ISO 9001:2000
Applications Industrial
Color Blue, Yellow
Latex Free No
Material Neoprene, Latex
Powdered Powder Free
Product Type Neoprene Gloves
Size 2X-Large
Thickness (English) 28 mil
Thickness (Metric) 0.71 mm
For Use With (Application) Petrochemical industry, janitorial, general maintenance, photo finishing
Glove Cuff Style Straight
Liner Material Flock
Lining Lined
Length (English) 12 in.
Length (Metric) 30.5 cm
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.