Learn More
PIP™ Bolle's Safety Override Protective Eyewear
Catalog No. 19036823 Shop All Protective Industrial Products Inc. Products
Change view
Click to view available options
Anti-scratch, anti-fog, clear PC lens
Catalog No.
19-036-823 Anti-scratch, anti-fog, clear PC lens
1 options
This item is not returnable. View return policy
Catalog No. 19-036-823 Supplier PIP™ Supplier No. 253OR40054
This item is not returnable. View return policy
Fits over prescription eyewear to provide high impact protection
Molded cheek/brow guard and side shield for optimum protection.
  • High impact 2.0mm polycarbonate lens
  • Anti-fog/anti-scratch coating on both sides
  • Optimal fit and comfort
  • With adjustable nylon temples
  • Single piece lens with distortion free viewing and superior optical quality
  • 99.99% UVA and UVB light absorption
  • Weighs 43g
Assembly and production work, Light engineering, Cutting/Brazing/Torch work
Specifications
Lens Coating Anti-fog, Scratch-resistant
Lens Tint Clear
For Use With (Application) Assembly and production work; Light engineering; Cutting/Brazing/Torch work
Includes Soft pouch and adjustable cord
No. per Pack 1 Pair
Temple Style Adjustable
Weight (Metric) 43 g
Lens Material Polycarbonate