Learn More
PIP™ Bouton™ Chemical Goggles
Catalog No. 19086609 Shop All Protective Industrial Products Inc. Products
Change view
Click to view available options
Lens Coating:
Anti-fog, Scratch-resistant
Uncoated
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Lens Coating
19-086609 Uncoated
19-823-725 Anti-fog, Scratch-resistant
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Catalog No. 19-086609 Supplier PIP™ Supplier No. 2484401300
Add to Cart
Add to Cart
This item is not returnable. View return policy
Four indirect vents help minimize fogging while keeping chemicals out
  • Polycarbonate lenses provide both impact and chemical splash protection
  • Fit comfortably over most prescription glasses
  • Flexible vinyl nose bridge adds comfort
Compliance
Meet ANSI™ Z87.1-2010 standards.
Specifications
Lens Coating Uncoated
Certifications/Compliance ANSI Z87.1
Lens Tint None
Product Type Indirect Vent Safety Goggles
For Use With (Application) General manufacturing; Anywhere eye protection is needed
Headband Material Elastic
Quantity 1/Polybag; 144/Cs.
Ventilation Details Indirect ventilation
Weight (Metric) 200 g
Material Vinyl
Disposable Reusable
Lens Material Polycarbonate
Frame Material Vinyl
UV Protection 99.99% UVA and UVB light absorption
Frame Type Indirect Vented
Show More Show Less
Adhere to
Safety Regulations and Best Practices
Take advantage of a range of Safety Services from our trusted suppliers to help ensure the safety and compliance of your people, processes, products, and facilities. Services include assessments, training, equipment maintenance, safety programs, and more.