Learn More
PIP™ Bouton™ Chemical Goggles
Catalog No. 19823725 Shop All Protective Industrial Products Inc. Products
Change view
Click to view available options
Lens Coating:
Anti-fog, Scratch-resistant
Uncoated
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Lens Coating
19-823-725 Anti-fog, Scratch-resistant
19-086609 Uncoated
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Catalog No. 19-823-725 Supplier PIP™ Supplier No. 2484401400
  • $1,932.00
    $1,168.65 / Case of 144
    Save $763.35
    40% Off The following deviations and disclosures may produce prices below stated discounts but will not trigger price reductions: Price shown reflects a temporary, promotional price reduction. No other purchase or commercial obligation required to receive the reduced price. Price may change at any time. Promotional price valid on web orders only. Your contract pricing may differ. Other exclusions apply.
Add to Cart
Add to Cart
This item is not returnable. View return policy
Four indirect vents help minimize fogging while keeping chemicals out
  • Polycarbonate lenses provide both impact and chemical splash protection
  • Fit comfortably over most prescription glasses
  • Flexible vinyl nose bridge adds comfort
Compliance
Meet ANSI™ Z87.1-2010 standards.
Specifications
Lens Coating Anti-fog, Scratch-resistant
Certifications/Compliance ANSI Z87.1-1989 standards
Lens Tint Clear
Product Type Chemical-resistant Goggles
Ventilation Details Four indirect vents
Material Vinyl
Disposable Reusable
Lens Material Polycarbonate
Frame Material Vinyl
Frame Type Indirect Vented
Adhere to
Safety Regulations and Best Practices
Take advantage of a range of Safety Services from our trusted suppliers to help ensure the safety and compliance of your people, processes, products, and facilities. Services include assessments, training, equipment maintenance, safety programs, and more.