Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

PIP™ Cabaret™ Cotton Dress Gloves with Raised Stitching - Open Cuff

Catalog No. 19816780 Shop All Protective Industrial Products Inc. Products
Change view
Click to view available options
Size:
Small
Large
X-Large
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Size
19-816-780 X-Large
19-816-779 Small
19-816-777 Large
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Catalog No. 19-816-780 Supplier PIP™ Supplier No. 130100WM/XL
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

Constructed in full fashion pattern with fourchettes and set-in thumbs for a tailored fit. Commonly used for formal occasions by police, firemen, waiters, marching bands, military, and pallbearers.

  • Three lines of raised stitching on back
  • Shirred elastic wrist
  • It is recommended to wash gloves in tempered water not exceeding 104°F (40°C)

Specifications

Construction String Knit
Cuff Material Shirred Elastic
Description Three lines of raised stitching on back of glove
Accelerator Free Yes
Applications Industrial
Coating Material Uncoated
Color White
Disposable Reusable
Exterior Finish Nap-out
Hand Specific
Material Cotton
Powdered Powder Free
Product Type Dress Gloves
Size X-Large
Sterility Non-sterile
Glove Cuff Color White
Glove Cuff Style Open
Hem Color White
Lining Seamless Lining
Palm Type Standard
Series 130-100WM
Thumb Style Keystone
Length (English) 10 in.
Length (Metric) 25.4 cm
Show More Show Less

Adhere to
Safety Regulations and Best Practices

Take advantage of a range of Safety Services from our trusted suppliers to help ensure the safety and compliance of your people, processes, products, and facilities. Services include assessments, training, equipment maintenance, safety programs, and more.

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.