Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

PIP™ CleanTeam™ Medium Weight Unhemmed Cotton Lisle Inspection Gloves

Catalog No. 19053688 Shop All Protective Industrial Products Inc. Products
Change view
Click to view available options
Construction:
Cut and Sewn
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Construction
19-053688 Cut and Sewn
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Catalog No. 19-053688 Supplier PIP™ Supplier No. 97520
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

Gloves are constructed in a two-piece, reversible pattern using 100% cotton fabric. Wear alone for use with inspection and electronics assembly, or as glove liners under disposable and unsupported liquid-proof gloves.

  • Constructed in a two piece, reversible pattern using 100% cotton fabric
  • Designed to minimize contamination of products from human hands
  • Can be used as liners under disposable and unsupported liquid proof gloves
  • Made from component materials which comply with Federal regulations for food contact: 21CFR, 170-99
  • Cut-and-Sewn construction
  • Unhemmed cuff
  • Silicone-free

Specifications

Certifications/Compliance 21 CFR 170-199
Construction Cut and Sewn
Coating Material Uncoated
Color White
Disposable Limited-use
Hand Ambidextrous
Latex Free Yes
Material Cotton
Product Type Inspection Gloves
Size Large
Sterility Non-sterile
Style Reversible
For Use With (Application) Electronics Assembly, Inspection
Gender Men's
Glove Cuff Style Unhemmed
Series 97-520
Length (English) 8.7 in.
Length (Metric) 22 cm
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.