Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

PIP™ CleanTeam™ Economy Nylon Full-Fashion Men's Inspection Gloves

Catalog No. 19121215C Shop All Protective Industrial Products Inc. Products
Change view
Click to view available options
Size:
Medium
Large
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Size
19-121-215C Large
19-121-215B Medium
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Catalog No. 19-121-215C Supplier PIP™ Supplier No. 98720L/PRC

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

Gloves are constructed of low lint 40-denier nylon tricot fabric to absorb perspiration, with set-in thumb and fourchettes for a tailored fit.

  • Manufacturer-suggested ISO Class 7 (Class 10,000)
  • Economical choice for hand protection or glove liners
  • Made of low-lint, 40-denier tricot fabric that absorbs perspiration for a comfortable fit
  • Full-fashion pattern with set-in thumb and fourchettes for tailored fit
  • Color-coded hem for quick size identification
  • Ambidextrous

Specifications

Coating Material Uncoated
Color White
Grade Economy
Hand Ambidextrous
Material Nylon
Product Type Inspection Glove
Size Large
Hem Color Green
Series 98-720
Thumb Style Keystone
Length (English) 9 in.
Length (Metric) 23 cm
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.