Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

PIP™ Seamless Knit Cotton/Polyester Gloves

Catalog No. 19149625 Shop All Protective Industrial Products Inc. Products
Change view
Click to view available options
Size:
Small
Large
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Size
19-149-625 Large
19-149-624 Small
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Catalog No. 19-149-625 Supplier PIP™ Supplier No. 35C115/L
This item is not returnable. View return policy

General purpose gloves are constructed from a seamless cotton/polyester blend liner for comfort and dexterity.

  • Seamless cotton/polyester knit shell is comfortable and flexible
  • Knit wrist helps prevent dirt and debris from entering the glove
  • Reversible design allows replacing one glove at a time, rather than pairs
  • Can be used as a glove liner
  • Small size fits ladies well
  • Made from component materials which comply with Federal regulations for food contact, 21 CFR, 170-199

Specifications

Certifications/Compliance 21 CFR, 170-199
Construction Seamless Knit
Coating Material Uncoated
Color Natural
Hand Specific
Material Polycotton
Product Type Seamless Knit Gloves
Size Large
For Use With (Application) Warehousing, Parts Handling, Assembly, and Food Processing
Glove Cuff Style Continuous Knit
Series 35-C115
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.